missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Phosphate, Disodium Salt Hexahydrate, MP Biomedicals

Catalog No. MP021008785
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021008785 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021008785 Supplier MP Biomedicals Inc Supplier No. 02100878.5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • p-nitrophenol is suitable for use as a substrate for alkaline and acid phosphatase

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium 4-nitrophenyl phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point >300°C (literature)
Boiling Point 457.84°C (760 mmHg(literature))
Linear Formula C6H4NO6PNa2 . 6H2O
Quantity 500 mg
Solubility Information Soluble in water.
Formula Weight 371.14
Physical Form Powder
Chemical Name or Material p-NITROPHENYL PHOSPHATE, DISODIUM SALT HEXAHYDRATE
Recommended Storage -20°C.