missing translation for 'onlineSavingsMsg'
Learn More

p-Nitro Blue Tetrazolium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog No. ICN19399983
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19399983 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19399983 Supplier MP Biomedicals Inc Supplier No. 0219399983

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.644
MDL Number MFCD00012159
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Color Yellow
Assay Percent Range 0.981
Quantity 250 mg
Formula Weight 817.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material p-Nitro Blue Tetrazolium Chloride, For HPLC analysis
EINECSNumber 206-067-4
Recommended Storage Store at +4°C.