missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxybenzoic Acid, Reagent, 99%, Spectrum™ Chemical

Catalog No. 18605194
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605194 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18605194 Supplier Spectrum Chemical Mfg Cor Supplier No. H1083100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

H1083, 99-96-7, HOC6H4COOH

Spectrum™ Chemical p-Hydroxybenzoic Acid, Reagent is also known as 4-Hydroxybenzoic acid and para-Hydroxybenzoic acid. It is mostly known as the basis for the preparation of its esters which are parabens. This is used in ophthalmic solutions and as a preservative in certain cosmetics. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

CAS Max % 1
Melting Point 213°C to 219°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99%
Grade Reagent
Physical Form Crystalline Powder or Lumps