missing translation for 'onlineSavingsMsg'
Learn More

Solution p-Diméthylaminobenzaldéhyde, pour hydrazine, nickel, certifiée, 1,8%, LabChem™

Numéro de catalogue. LC135901
Modifier l'affichage
Click to view available options
Quantité:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
LC135901 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC135901 Fournisseur LabChem Code fournisseur LC135901

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 100-10-7 , 67-56-1 , 7647-01-0
Formule moléculaire CH4O
Poids moléculaire (g/mol) 32.04
Numéro MDL MFCD00004595
Clé InChI OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonyme 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
Nom de l’IUPAC Méthanol
SOURIRES CO

Spécifications

Nom ou substance chimique p-Dimethylaminobenzaldehyde Solution
Nom Remarque Pour l’hydrazine, le nickel
Couleur Jaune
CAS % maximal 1.79, 89.28, 8.93
Formule linéaire C9H11NO
Numéro UN UN2924
Renseignements sur la solubilité Soluble dans l’eau
Poids de formule 149.19
Pourcentage de pureté 1.8%
Niveau Certifié
Désignation Test de réussite
Emballage Verre ambré
Aptitude Test de réussite
Renseignements sur la décomposition Chlorure d’hydrogène; Monoxyde de carbone; Le dioxyde de carbone; Ammoniac gazeux
Quantité 500 mL
Forme physique Liquide
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs (central nervous system, optic nerve).
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Température ambiante