missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine Hydrochloride 99.0+%, TCI America™

Catalog No. A049025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A049025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A049025G Supplier TCI America Supplier No. A049025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20265-97-8
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00036388
InChI Key VQYJLACQFYZHCO-UHFFFAOYSA-N
Synonym p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride
PubChem CID 2734956
IUPAC Name 4-methoxyaniline;hydrochloride
SMILES COC1=CC=C(C=C1)N.Cl

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 159.61
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine Hydrochloride
RTECSNumber BZ6600000
TSCA Yes