missing translation for 'onlineSavingsMsg'
Learn More

p-Acetanisidide 98.0+%, TCI America™

Catalog No. A001925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001925G 25 g
A0019100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A001925G Supplier TCI America Supplier No. A001925G
Only null left

Chemical Identifiers

CAS 51-66-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00014963
InChI Key XVAIDCNLVLTVFM-UHFFFAOYSA-N
Synonym n-4-methoxyphenyl acetamide, 4'-methoxyacetanilide, p-methoxyacetanilide, p-acetanisidide, p-acetanisidine, methacetin, 4-methoxyacetanilide, aceto-p-anisidide, n-acetyl-p-anisidine, acetamide, n-4-methoxyphenyl
PubChem CID 5827
IUPAC Name N-(4-methoxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)OC

Specifications

Melting Point 130°C
Color White
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Acetanisidide
EINECSNumber (3)-0690
RTECSNumber AE8290000
TSCA Yes