missing translation for 'onlineSavingsMsg'
Learn More

Oxyclozanide, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022886
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022886 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022886 Supplier Merck Emd Millipore Supplier No. BP273

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 2277-92-1
Molecular Formula C13H6Cl5NO3
Linear Formula C13H6Cl5NO3
MDL Number MFCD00864507
Quantity 200 mg
Synonym 2,3,5-Trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide; 3,3',5,5',6-Pentachloro-2'-hydroxysalicylanilide
Formula Weight 401.46
Grade BP