missing translation for 'onlineSavingsMsg'
Learn More

Oxyclozanide, 98%, Thermo Scientific Chemicals

Catalog No. AC460881000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460881000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460881000 Supplier Thermo Scientific Chemicals Supplier No. 460881000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2277-92-1
Molecular Formula C13H6Cl5NO3
Molecular Weight (g/mol) 401.45
MDL Number MFCD00864507
InChI Key JYWIYHUXVMAGLG-UHFFFAOYSA-N
Synonym oxyclozanide, oxiclozanidum, zanil, oxyclozanid, zanilox, diplin, 2,3,5-trichloro-n-3,5-dichloro-2-hydroxyphenyl-6-hydroxybenzamide, oxyclozanide inn:ban, oxyclozanidum inn-latin, oxiclozanida inn-spanish
PubChem CID 16779
IUPAC Name 2,3,5-trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide
SMILES OC1=C(C(=O)NC2=CC(Cl)=CC(Cl)=C2O)C(Cl)=C(Cl)C=C1Cl

Specifications

Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula Cl3C6H(OH)C(=O)NHC6H2(OH)Cl2
Quantity 100 mg
Formula Weight 401.46
Percent Purity 98%
Chemical Name or Material Oxyclozanide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid release to the environment.
EINECSNumber 218-904-

RUO – Research Use Only