Learn More
Thermo Scientific™ Oxoid™ V Factor Discs
Catalog No. OXDD0004T
Change view
Click to view available options
Quantity:
50 Disks/Cartridge
Catalog No. Quantity
OXDD0004T 50 Disks/Cartridge
1 options
Catalog No. OXDD0004T Supplier Thermo Scientific™ Supplier No. DD0004T
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Ordering Alerts:
50 Discs/Cartridge
Specifications
Content And Storage -10°C to -20°C
Description V Factor Discs
Detectable Analytes Haemophilus species
Type X Factor Disc
Packaging Type 1 Cart./Pk.
Quantity 50 Disks/Cartridge