missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thioglycollate Medium USP (Dehydrated)

Numéro de catalogue. p-4524942
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXCM0173B 500 g
OXCM0173T 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. OXCM0173B Fournisseur Thermo Scientific™ Code fournisseur CM0173B
Il en reste null

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with this medium.

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with Thermo Scientific™ Oxoid™ Thioglycollate Medium USP (Dehydrated). The medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests and conforms to formulations detailed in the British and European Pharmacopoeia 2,3,4.

Thioglycollate Medium is suitable for the cultivation of both aerobic and anaerobic organisms. Sodium thioglycollate acts as a reducing agent and neutralizes the toxic effects of mercurial compounds used as preservatives in solutions for injection, etc.

  • Well-buffered medium ensures that acid or alkaline inocula produce negligible alteration in the reaction of the medium
  • Suitable for cultivation of both aerobic and anaerobic organisms.

Not all products are available for sale in all territories. Please inquire.

Spécifications

Description Thioglycollate Medium, USP
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 16.9L medium

For Laboratory Use Only