missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Streptococcal Grouping Disposable Reaction Card

Catalog No. OXDR0500G
Change view
Click to view available options
Quantity:
50 Cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDR0500G 50 Cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDR0500G Supplier Thermo Scientific™ Supplier No. DR0500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxoid Streptococcal Grouping Disposable Reaction Card is used with Streptococcal Grouping Reagents.

A latex agglutination test for the identification of streptococcal groups A, B, C, D, F, and G.

Specifications

Content And Storage Freeze-dried 2° to 25°C and reconstituted at 2° to 8°C
Description Streptococcal Grouping Disposable Reaction Card
Type Reaction Card for Streptococcus Testing
Organism Group Group A, B, C, D, F and G
Quantity 50 Cards