missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Perfringens SFP Supplement

Catalog No. OXSR0093E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0093E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0093E Supplier Thermo Scientific™ Supplier No. SR0093E

Oxoid Perfringens SFP Supplement is for the selective isolation of Clostridium perfringens.

Oxoid Perfringens SFP Supplement is for the selective isolation of Clostridium perfringens.

  • Add to Perfringens Agar Base, TSC, Part No. CM0587B
  • Each vial supplements 500mL of medium

Specifications

Description Perfringens SFP Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Perfringens