missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MYP Agar (Dehydrated)

Numéro de catalogue. OXCM0929B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXCM0929B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. OXCM0929B Fournisseur Thermo Scientific™ Code fournisseur CM0929B
Il en reste null

Enumerate Bacillus cereus in food samples with this selective and differential medium.

Enumerate Bacillus cereus in food samples with selective and differential medium Thermo Scientific™ Oxoid™ MYP Agar (Dehydrated). The diagnostic features of the medium rely upon the failure of Bacillus cereus to utilize mannitol and the ability of most strains to produce phospholipase C, resulting in an opaque halo around the colonies.

Bacillus cereus has been recognized as a causative agent of food poisoning since the 1950s. Many early isolation techniques used blood agar, relying on hemolysis and colony morphology, followed by a range of tests for confirmation. MYP Agar has proven to be very effective for detecting B. cereus even with very low numbers.

  • Selective: due to the presence of Polymyxin B the medium inhibits most Gram-negative bacteria.
  • Easy-to-read: easy to differentiate Bacillus cereus (opaque halo zone of precipitation around the colonies) from other Gram-positive organisms (no zones of inhibition).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Spécifications

Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, FDA, ISO, NMKL.
Description MYP Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.6L medium

For Laboratory Use Only