missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Escherichia coli O157 Latex Test

Catalog No. OXDR0620M
Change view
Click to view available options
Quantity:
100 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDR0620M 100 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDR0620M Supplier Thermo Scientific™ Supplier No. DR0620M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly confirm E. coli serogroup O157 from colonies cultured on plated media by latex agglutination.

A latex agglutination test for the identification of E. coli serogroup O157. Certain strains of Escherichia coli have been implicated in cases of haemorrhagic colitis (HC) and haemolytic ureamic syndrome (HUS). It has been shown that these strains produce a vero-cytotoxin (VT). The E coli serotype most frequently isolated from HC and HUScases is O157:H7. Isolation of this serotype from a diarrhoeal stool particularly with the presence of blood is indicative of a vero-cytotoxin producing strain1, 2, 3, 4, 5, 6, 7 ,8. The Oxoid E coli O157 latex test will demonstrate by slide agglutination E. coli strains possessing the O157 serogroup antigen. The test is best used in conjunction with Sorbitol MacConkey Agar (Oxoid CM0813). E. coli O157:H7 strains do not ferment Sorbitol and therefore give colourless colonies on this medium. The majority of E coli isolates do ferment Sorbitol and give characteristic pink colonies. Sorbitol MacConkey Agar should be used as the primary screen. Non-sorbitol fermenting colonies can then be tested with the latex reagents, to determine whether the isolate belongs to the O157 serogroup and therefore a potential VT- producing strain.

Use this dry latex test designed to demonstrate by slide agglutination E. coli strains possessing the O157 serogroup antigen.

The test is best used in conjunction with Sorbitol MacConkey Agar. E. coli O157:H7 strains do not ferment sorbitol and, therefore, give colorless colonies on this medium. The majority of E. coli isolates do ferment sorbitol and give characteristic pink colonies. Sorbitol MacConkey Agar should be used as the primary screen. Non-sorbitol fermenting colonies can then be tested with the latex reagents to determine whether the isolate belongs to the O157 serogroup and, therefore, a potential verotoxin-producing strain.

  • Pre-dispensed reagents eliminate extra steps for faster time to result
  • Room temperature storage for up to two years simplifies inventory
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards for convenience

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 2°C to 8°C
Description E. coli O157 Latex Test
Detectable Analytes E. coli
Sample Type E. coli Colonies from Plate
Type E. coli Testing Kit
Quantity 100 Tests/Kit