missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 1 Latex Agglutination Test

Catalog No. OXDR0200M
Change view
Click to view available options
Quantity:
60 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXDR0200M 60 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDR0200M Supplier Thermo Scientific™ Supplier No. DR0200M

missing translation for 'validMainframeMsg'

Rapidly confirm Legionella pneumophila serogroup 1 from culture by latex agglutination.

The DrySpot Legionella Latex Test is a latex agglutination test for the identification of Legionella species grown on plate media from patients with suspected legionellosis or from environmental sources. The DrySpot Legionella Latex Test allows separate identification of Legionella pneumophila serogroup 1, serogroups 2–14 and seven other Legionella species that have been implicated in human disease.

Use this dry latex agglutination test for the identification of Legionella pneumophila serogroup 1 isolated either from patients with suspected legionellosis or from environmental sources.

Blue latex particles sensitised with specific rabbit antibody are dried onto the test card. These react with Legionella pneumophila serogroup 1 antigen to form agglutination in the test reaction area.

Blue latex particles sensitised with non-reactive rabbit globulins are dried onto the test card to create the control reaction area.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 2°C to 25°C
Description DrySpot Legionella Pneumophila Serogroup 1 Latex Test
Detectable Analytes Legionella
Sample Type Human Stool
Type Legionella Latex Test Kit
Includes 20 reaction cards (3 tests/card), positive control, negative control, phosphate buffered saline, mixing paddles, pouch clips
Quantity 60 Tests/Kit