missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Azide Dextrose Broth (Dehydrated)

Numéro de catalogue. OXCM0868B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXCM0868B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. OXCM0868B Fournisseur Thermo Scientific™ Code fournisseur CM0868B
Il en reste null

Detect enterococci in water and sewage samples with this selective medium.

Detect enterococci in water and sewage samples with Thermo Scientific™ Oxoid™ Azide Dextrose Broth (Dehydrated). A blend of peptone and glucose in the medium renders Azide Dextrose Broth highly nutritious, and sodium chloride maintains the osmotic equilibrium. Sodium azide acts as an inhibitor of Gram-negative organisms1,2,3 and the concentration of sodium azide used in the medium provides optimum protection for the enterococci while largely suppressing the Gram-negative flora. The phosphate buffer system controls pH.

Enterococci are part of the normal intestinal flora of human and animals. They have been long recognized as important human pathogens and are becoming increasingly so. The presence of enterococci serves as an indicator of fecal contamination. Enterococci are better indicators than Escherichia coli of sewage pollution in chlorinated waters because they have a greater resistance to chlorine.

Azide Dextrose broth is recommended for the detection of enterococci in water and sewage4.

  • Selective: sodium azinde in the medium acts as an inhibitor of Gram-negative organisms while providing optimum protection to enterococci.
  • Recommended for the quantitative determination of enterococci in water, sewage, foods and other materials suspected of contamination with sewage1.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Spécifications

Description Azide Dextrose Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 14.0L medium

For Laboratory Use Only