missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ AnaeroJar™ Clip

Catalog No. OXAG0031A
Change view
Click to view available options
Quantity:
2/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXAG0031A 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXAG0031A Supplier Thermo Scientific™ Supplier No. AG0031A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxoid AnaeroJar Clip has two self-venting clips.

Oxoid AnaeroJar Clip has two self-venting clips.

For use with AnaeroJar 2.5L, Part No. AG0025A

Includes:
2 self-venting clips.

Specifications

Type AnaeroJar Clip
Format Self-venting Clip
Includes 2 self-venting clips
Product Line Thermo Scientific Oxoid
Quantity 2/Pk.