missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Oxcarbazepine

Catalog No. 386410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
386410 10 mg
386450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 386410 Supplier Tocris Bioscience™ Supplier No. 3864/10

Anticonvulsant; inhibits Na+ channel activity

Anticonvulsant; protects mice and rats against generalized tonic-clonic seizures induced by electroshock. Thought to act via inhibition of sodium channel activity.

Chemical Identifiers

CAS 28721-07-5
Molecular Formula C15H12N2O2

Specifications

Quantity 10 mg
Formula Weight 252.27
Purity >98%
Molecular Weight (g/mol) 252.273
Chemical Name or Material Oxcarbazepine
Recommended Storage Store at Room Temperature