missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxantel pamoate, MilliporeSigma™ Supelco™
Description
Pharmaceutique (petite molécule)
Spécifications
Spécifications
| Packaging | Bouteille en polyéthylène |
| CAS | 68813-55-8 |
| Physical Form | Sympa |
| Linear Formula | C13H16N2 O · C23H16O6 |
| MDL Number | MFCD19105662 |
| Synonym | 1-Methyl-2-(3-hydroxyphenylethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4 ′-methylenebis(3-hydroxy-2-naphthoic acid); Oxantel embonate |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 604.65 |
| Grade | Norme analytique |