missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl chloride, 2M soln. in dichloromethane

Catalog No. AAH31617AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31617AP 500 mL
AAH31617AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31617AP Supplier Thermo Scientific Chemicals Supplier No. H31617AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-37-8 | C2Cl2O2 | 126.92 g/mol

Reagent used in organic synthesis. Unncatalyzed reaction of silyl ketene acetals with oxalyl chloride yeilds symmetrical pulvinic acids. Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride. Oxalyl chloride was reportedly used in the first synthesis of dioxane tetraketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used in organic synthesis. Unncatalyzed reaction of silyl ketene acetals with oxalyl chloride yeilds symmetrical pulvinic acids. Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride. Oxalyl chloride was reportedly used in the first synthesis of dioxane tetraketone.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, light, bases, alkali metals, alcohols, dimethyl formamide.

Chemical Identifiers

CAS 79-37-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.92
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl

Specifications

Density 1.335
Quantity 500 mL
UN Number UN2922
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 126.93
Concentration or Composition (by Analyte or Components) 2M soln. in dichloromethane
Chemical Name or Material Oxalyl chloride
Hazard Category H302+H332-H314-H350
Hazard Statement GHS H Statement
H331-H314-H318-H351
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 201-200-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only