missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid TS, 6.3% (w/v), Ricca Chemical

Catalog No. 546616
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
546616 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 546616 Supplier Ricca Chemical Company Supplier No. 546616
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1.3 (0.1M)
Melting Point ∼0°C
Boiling Point ∼100°C
CAS Min % 92.0, 5.81
Color Colorless
CAS Max % 96.0, 6.05
Concentration 6.3% (w/v)
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Specific Gravity ∽1.02 to 1.05
Solubility Information Infinite
Percent Purity 6.3% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6.3% (w/v)
Chemical Name or Material Oxalic Acid, Test Solution
Show More Show Less