missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 0.100 N (N/10), 0.0500 M (M/20), Ricca Chemical

Catalog No. 5471116
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5471116 500 mL
5471132 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5471116 Supplier Ricca Chemical Company Supplier No. 5471.116

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

CAS Min % 97.0, 0.63
CAS Max % 100.0, 0.65
Grade Laboratory
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material OXALIC ACID STOCK SOLUTION