missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid , For Silica, Certified, 7.5% (w/v) ±0.5%, LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 92.5, 7.5 |
| Quantity | 1 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Traceability to NIST | Traceable to NIST |
| Linear Formula | HO2CCO2H |
| Solubility Information | Soluble in water |
| Formula Weight | 90.04 |
| Percent Purity | 7.5% ±0.5% (w/v) |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |