missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid , For Silica, 5% (w/v), LabChem™

Catalog No. LC180501
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC180501 500 mL
LC180502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC180501 Supplier LabChem Supplier No. LC180501

Chemical Identifiers

CAS 6153-56-6 , 7732-18-5
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Color Colorless
CAS Max % 95, 5
Linear Formula HO2CCO2H
Solubility Information Soluble in water
Formula Weight 90.04
Percent Purity 5% (w/v)
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Chemical Name or Material Oxalic Acid, For Silica
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature