missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCO0200500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCO0200500 500 g Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCO0200500 Supplier Ricca Chemical Company Supplier No. RDCO0200500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 500 g
Packaging Poly Wide Mouth Bottle
SMILES C(=O)(C(=O)O)O.O.O
IUPAC Name oxalic acid;dihydrate
Molecular Weight (g/mol) 126.064
Percent Purity 99.5 to 102.5%
Grade ACS
Chemical Name or Material Oxalic Acid, Dihydrate
ShelfLife 60 months