missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid , Dihydrate, ACS Grade, 99.5 to 102.05%, LabChem™

Catalog No. LC180401
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC180401 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC180401 Supplier LabChem Supplier No. LC180401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Melting Point 101°C
Color White
CAS Max % 100
Density 1,653kg/m3
Quantity 500 g
Packaging Poly Bottle
Physical Form Crystalline Powder
Specific Gravity 1653kg/m3
Linear Formula HO2CCO2H
UN Number UN3261
Solubility Information Soluble in water
Formula Weight 90.04
Percent Purity 99.5 to 102.05%
Grade ACS
Assay as dihydrate
Chemical Name or Material Oxalic Acid, Dihydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 205-634-3
Recommended Storage Room Temperature