missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 7.5% (w/v), Ricca Chemical

Catalog No. 546716
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
546716 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 546716 Supplier Ricca Chemical Company Supplier No. 546716

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Percent Purity 7.5% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 7.5% (w/v) Aqueous Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Oxalic Acid