missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 5% (w/v), Ricca Chemical

Catalog No. 5465516
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5465516 500 mL Natural Poly Bottle
5465532 1 L Natural Poly Bottle
R54655004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5465516 Supplier Ricca Chemical Company Supplier No. 5465.516

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6

Specifications

Synonym dihydrogen oxide, dihydrogen monoxide
Percent Purity 5% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5% (w/v) Aqueous Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Oxalic Acid