missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid, 10% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Quantity | 20 L |
| Odor | Odorless |
| Packaging | Cubitainer |
| Physical Form | Liquid |
| Solubility Information | Solubility in Water: Infinite |
| Percent Purity | 10% w/v |
| Grade | Laboratory |
| Chemical Name or Material | Oxalic Acid, 10% (w/v) |