Recombinant Protein;50UG;Human OX40 Ligand, His Tag (active trimer) (MALS verified) (OXL-H52Q8) is expressed from human 293 cells (HEK293). It contains AA Gln 51 - Leu 183 (Accession # NP_003317).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.9 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human OX40, Fc,Avitag (Cat. No. OX0-H82F7) at 2 g/mL (100 L/well) on streptavidin precoated (0.5 g/well) plate, can bind Human OX40 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. OXL-H52Q8) with a linear range of 5-39 ng/mL (QC tested).;Immobilized Human OX40 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. OXL-H52Q8) at 2 g/mL (100 L/well) can bind Biotinylated Human OX40, Fc,Avitag (Cat. No. OX0-H82F7) with a linear range of 10-78 ng/mL (Routinely tested).;FACS assay shows that Human OX40 Ligand, His Tag (active trimer) (MALS verified) (Cat. N