Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human OX40, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Numéro de catalogue. 501659158
Encompass
Recombinant Protein;Biotinylated Human OX40, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human OX40, Fc Tag, Avi Tag (OX0-H82F7) is expressed from human HEK293 cells. It contains AA Leu 29 - Ala 216 (Accession # NP_003318).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 48.6 kDa. The protein migrates as 66 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) at 5 g/mL (100 l/well) can bind Biotinylated Human OX40, Fc Tag, Avi Tag (Cat. No. OX0-H82F7) with a linear range of 0.01-0.3 g/mL (QC tested).;FACS assay shows that recombinant Biotinylated Human OX40, Fc Tag, Avi Tag (Cat. No. OX0-H82F7) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 is 0.1 g/ml (Routinely tested).
Numéro de catalogue. 501659158 Fournisseur ACROBiosystems Inc Code fournisseur OX0H82F725UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Biotinylated Human OX40, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human OX40, Fc Tag, Avi Tag (OX0-H82F7) is expressed from human HEK293 cells. It contains AA Leu 29 - Ala 216 (Accession # NP_003318).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 48.6 kDa. The protein migrates as 66 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) at 5 g/mL (100 l/well) can bind Biotinylated Human OX40, Fc Tag, Avi Tag (Cat. No. OX0-H82F7) with a linear range of 0.01-0.3 g/mL (QC tested).;FACS assay shows that recombinant Biotinylated Human OX40, Fc Tag, Avi Tag (Cat. No. OX0-H82F7) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 is 0.1 g/ml (Routinely tested).