missing translation for 'onlineSavingsMsg'
Learn More

Osmium powder, -20 mesh, 99.95% (metals basis)

Catalog No. AA3999806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3999806 5 g
AA3999803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3999806 Supplier Thermo Scientific Chemicals Supplier No. 03999806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-04-2 | Os | 190.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4-2-7440
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium

Specifications

Quantity 5 g
Mesh Size -20 Mesh
Assay Percent Range 99.95% (metals basis)
Molecular Formula Pd
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES [Pd]
Molecular Weight (g/mol) 106.42
Chemical Name or Material Osmium powder
Hazard Category H228-H315-H318-H335
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-114-0
RTECSNumber RN1100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only