missing translation for 'onlineSavingsMsg'
Learn More

Precision Systems Osmette™ A Osmometer

Numéro de catalogue. 128275
Modifier l'affichage
Click to view available options
Display Type:
Digital
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Display Type
128275 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 128275 Fournisseur Precision Systems Code fournisseur 5002

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette A Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.

Easy to Use

  • Add sample tube to well, lower operating head to insert wire and thermistor temperature probe into the sample
  • Freezing point is detected by the probe, with accuracy of ±0.001°C
  • Digital display shows results directly in mOsm/kg H2O
  • Entire cycle takes between one-half and one minute per sample
Speedy and convenient
  • Fast cooldown, completely zero-stable meter
  • Reach operating temperature in <15 min. vs. 40 to 60 min. for other cryoscopic devices
  • Eliminate Dewar flasks, ice baths, calculations
  • Generate precise results in just 50 to 70 sec.; process 40 to 60 samples per hour
  • Minimal training; simple two-step calibration takes less than five minutes
  • All models hold calibration, eliminating daily calibration

Osmette A, Model 5002

  • Automatic Operation
  • Small 2 oz. (59mL) bath allows faster results with better control
  • Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control
  • Clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids

  1. Reproducibility, 1 S.D. (one standard deviation).
  2. With liquid bath.
  3. Also available: Cryette™ Cryoscopes. Model 5006 (Cat. No. 12-672) determines added water in milk; Models 5008 and 5009 (Cat. Nos. 12-672-1 and 12-672-2) determine molecular weight of solutions such as petroleum fractions and core samples. For information, contact your Fisher Scientific Customer Service Representative.

Spécifications

Display Type Digital
Frequency 50/60 Hz
Voltage 115 V
Cooling Agent Thermoelectric Refrigerator
Quantity Each
Product Type Osmometer
Model 5002
For Use With (Application) Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control, clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids
Height (English) 10 in.
Height (Metric) 25 cm
Length (English) 8 in.
Measurement Time <1 min.
Length (Metric) 20 cm
Operation Automatic
Precision For 2mL sample: <1000mOsm/kg: ±1mOsm/kgH2O, >1000mOsm/kgH2O: ±0.2%. For 0.2mL sample: <500mOsm/kg: ±2mOsm/kgH2O, >500mOsm/kgH2O: ±0.5%
Range 0 to 3000 mOsm/kg H2O
Sample Size 2 or 0.2 mL
Width (Metric) 38 cm
Width (English) 15 in.
Cool-Down Time 10 to 15 min.
Dimensions (L x W x H) 8 x 15 x 10 in. (20 x 38 x 25 cm)
Electrical Requirements 115 V 50/60 Hz
Afficher plus de résultats Afficher moins de résultats