missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard 600 mV vs. Ag/AgCl, Reagents

ORP Standard 600 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CO00050020F

 View more versions of this product

Catalog No. 01-334-581


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

ORP Standard 600 mV vs. Ag/AgCl
7732-18-5 , 7783-85-9 , 7664-93-9
Liquid
Amber
Oxidation Reduction Potential Standard
Completely miscible
Reagent
Cubitainer
1.0286
Redox
100°C
Redox Meter
20 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions