missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard 600 mV vs. Ag/AgCl, Reagents

Catalog No. p-200021670
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334582 1 L
01334581 20 L
01334580 4 L
01334579 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334582 Supplier Labchem Inc Supplier No. CO0005001C
Only null left

ORP Standard 600 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard 600 mV vs. Ag/AgCl
Packaging Glass Bottle
CAS 7732-18-5, 7783-85-9, 7664-93-9
Specific Gravity 1.0286
Physical Form Liquid
Technique Redox
Color Amber
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less