missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, +600 mV, Ricca Chemical

Catalog No. 546464
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
546464 120 mL Amber Glass Bottle
5464616 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 546464 Supplier Ricca Chemical Company Supplier No. 5464.64

Chemical Identifiers

CAS 7783-85-9 , 7783-83-7 , 7664-93-9 , 7732-18-5
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Packaging Amber Glass Bottle
CAS Min % 92.0, 4.43, 1.69, 0.02
CAS Max % 96.0, 4.61, 1.76, 0.02
Physical Form Liquid
Color Amber
Quantity 120 mL
Concentration or Composition (by Analyte or Components) 4M KCl sat. with AgCl filling solution
Chemical Name or Material Oxidation-Reduction Potential Standard, +600mV vs. Ag/AgCl
ShelfLife 9 months