missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, 475 mV, Reagents

Catalog No. 01334572
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334572 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334572 Supplier Labchem Inc Supplier No. CO005000500B

ORP Standard, 475 mV, Reagent, use for Redox. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard, 475 mV
Packaging Opaque HDPE
CAS 7732-18-5, 7664-93-9, 7783-85-9
Specific Gravity 1.0846
Physical Form Liquid
Technique Redox
Color Yellow/Green
pH <1
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 500 mL
Grade Reagent
Show More Show Less