missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, +400 mV, Ricca Chemical

Catalog No. R5464540500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5464540500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5464540500 Supplier Ricca Chemical Company Supplier No. R5464540500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-85-9 , 7783-83-7 , 7664-93-9 , 7732-18-5
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Packaging Amber Glass Bottle
CAS Min % 86.78, 9.35, 3.63, 0.24
CAS Max % 86.78, 9.35, 3.63, 0.24
Density 1.08
Physical Form Liquid
Color Green to Tan/Yellow
Boiling Point Approximately 100°C
For Use With (Equipment) ORP Meter
Solubility Miscible with water
Solution Type Calibration Standard
Type Redox/ORP Electrode Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material ORP Standard, +400 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution)
Show More Show Less
ShelfLife 12 months
Recommended Storage Refridgerated Corrosive