missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, 200mV, Certified, LabChem™

Catalog No. lc180101
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC180101 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC180101 Supplier LabChem Supplier No. LC180101
Only null left

Chemical Identifiers

CAS 14459-95-1 , 13746-66-2 , 7447-40-7 , 7732-18-5
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Packaging Amber Poly
CAS Max % 98.64, 0.53, 0.75, 0.08
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Linear Formula K3Fe(CN)6
Color Yellow
Solubility Soluble in water
Quantity 500 mL
Formula Weight 329.26
Grade Certified
Chemical Name or Material ORP Standard, 200mV
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature