Learn More
Ornidazole, 99%
Description
Ornidazole is a drug that cures some protozoan infections. It has been investigated for use in Crohn's disease after bowel resection. It is indicated in treatment of mixed amoebiasis, mixed amoebic dysentery, mixed giardiasis, trichomoniasis, bacterial vaginosis, Sexually transmitted diseases, infections of gynaecology, lower respiratory tract infections, ENT, surgical and dental infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in ethanol at 50mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Spécifications
Spécifications
| Melting Point | 84°C to 86°C |
| Color | White to Yellow |
| Quantity | 5 g |
| Merck Index | 14,6872 |
| Solubility Information | Soluble in ethanol at 50mg/ml |
| Formula Weight | 219.63 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | Ornidazole |
Sécurité et manipulation
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 240-826-0 |
| RTECSNumber | NI5460000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only