Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Numéro de catalogue. p-4529778
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Numéro de catalogue. Connector Types
13645161 BNC (Waterproof)
1 options
Numéro de catalogue. 13645161 Fournisseur Thermo Scientific™ Code fournisseur 9682BNWP
Il en reste null
Measure lead ions in aqueous solutions quickly, simply, accurately and economically.
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
Features
Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap The lead electrode makes a sensitive endpoint detector for the titration of sulfate ions with lead perchlorate.
Solutions required:
Standard
948206 – 0.1 M lead standard, 475 mL bottle
948207 – 0.1 M sodium sulfate standard for sulfateanalysis with the lead ISE, 475 mL bottle
Electrode Fill Solution
900062 – Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included
900062 – Optimum Results B electrode filling solution, 60 mL 1 bottle
948201 – Polishing strip. Qty 2.
Warranty period
12 months from date of purchase
Spécifications
Connector Types BNC (Waterproof)
Description With waterproof BNC connector
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Lead Ion Selective Electrode, BNC waterproof connector