missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Double Junction Semi-Micro Spear KnipHe™ Glass Bodied Combination pH Electrode
Description
Thermo Scientific™ Orion™ Spear Tip Double Junction pH Electrodes provide reliable and accurate measurements of meat, cheese and sludge samples. These electrodes are best suited for piercing solid/semi-solid samples and measuring small volume samples.
Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.
Solutions required
Electrode fill solution
910008 3M KCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
910008 3M KCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Specifications
| Description | Orion 9121APWP Double Junction Replacement KNIpHE Combination pH electrode |
| Connector Types | BNC |
| Junction | Ceramic |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | Combination pH Electrode, Replacement |
| Filling Solution Type | 3M KCl |
| For Use With (Equipment) | Orion pH Meter |
| Internal Reference | Ag/AgCl Double Junction |
| Storage Requirements | Store electrode in Orion pH electrode storage solution 910001 |
| Show More |