missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS™ Refillable Epoxy Bodied Combination pH Electrode

Catalog No. 13641760
Change view
Click to view available options
Connector Types:
BNC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13641760 BNC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13641760 Supplier Thermo Scientific™ Supplier No. 815600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Confidently maneuver and measure in the field or on the plant floor with Thermo Scientific™Orion™ ROSS™ Refillable Epoxy Body Electrodes.

Thermo Scientific™ Orion™ ROSS™ Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in epoxy bodies for ruggedness and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.

Orion ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required

  • Electrode fill solution
  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase

Specifications

Description With BNC connector
Connector Types BNC
Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination pH Electrode
Drift <0.002pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter
Internal Reference ROSS
Precision 0.01
Temperature (English) Operating 32°F to 212°F
Show More Show Less