missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied Reference Electrode

Catalog No. 13299602
Change view
Click to view available options
Length (Metric):
120 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
13299602 120 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13299602 Supplier Thermo Scientific™ Supplier No. 800500U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform precise pH determinations for routine or research work.

This Reference Half-Cell Electrode is for use with ROSS Half-Cell pH Electrode 8101BNWP or 8411BN ROSS sodium half-cell electrodes.
 

Thermo Scientific™ Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 24 months from date of purchase

Specifications

Description 800500U ROSS Ultra Reference Electrode
Connector Types BNC (Waterproof)
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Reference Electrode
Electrode Reference half-cell glass body electrode; Pin tip connector
Filling Solution Type ROSS electrode fill solution
For Use With (Application) Precise pH determinations
Internal Reference ROSS
Precision 0.01
Length (Metric) 120 mm
Diameter (Metric) Body 120 mm
Diameter (Metric) Cap 16 mm
Show More Show Less