missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 100 ppm C, Ricca Chemical

Supplier:  Ricca Chemical Company R18460001C

 View more versions of this product

Catalog No. R18460001C


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Organic Carbon Standard
7732-18-5,7664-38-2,877-24-7
Liquid
Colorless
100°C
Standard
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Mixture
23676735
Amber Glass Bottle
Odorless
C8H5KO4
<2
Infinitely soluble in water
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
1 L
Laboratory

Chemical Identifiers

7732-18-5
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
C8H5KO4
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : General

SDS
Documents

Documents

Promotions

Promotions