missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 100 ppm C, Ricca Chemical
$214.63 - $229.19
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C8H5KO4 |
| InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
| PubChem CID | 23676735 |
| IUPAC Name | potassium;2-carboxybenzoate |
| SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R1846000250
|
Ricca Chemical Company
R1846000250C |
250 mL | Amber Glass Bottle |
Each for $214.63
|
|
||||
|
R18460001C
|
Ricca Chemical Company
R18460001C |
1 L | Amber Glass Bottle |
Each for $229.19
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate |
| C8H5KO4 | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Specifications
| Amber Glass Bottle | |
| 99.58,0.4,0.02 | |
| 1.001 | |
| C8H5KO4 | |
| < 2 | |
| 100°C | |
| Standard | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| 204.222 | |
| 23676735 | |
| Organic Carbon Standard, 100 ppm C |
| 7732-18-5,7664-38-2,877-24-7 | |
| 99.58,0.4,0.02 | |
| Liquid | |
| Colorless | |
| 0°C | |
| Miscible with water | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate | |
| 250 mL | |
| Laboratory |
Safety and Handling
Recommended Storage : General