Learn More
MilliporeSigma™ Supelco™ ORBO™ 609 Amberlite XAD-2 (20/50) 400/200 mg W,W,W separators
Catalog No. 111009109
Change view
Click to view available options
Diameter (Metric):
8 mm
Catalog No. Diameter (Metric)
111009109 8 mm
1 options
Catalog No. 111009109 Supplier MilliporeSigma™ Supelco™ Supplier No. 20051
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used for gas and vapor sampling
ORBO™ 609 Amberlite™ XAD™-2 (20/50), 400/200 mg sorbent tubes (W,W,W separators, O.D. x L 8 mm x 110 mm) contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Specifications
Description Bed A 400 mg/Bed B 200 mg
Material W,W,W Separators
Pore Size 20 to 50 Mesh
Quantity 50/Pk.
Length (Metric) 110 mm
Diameter (Metric) 8 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC