missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ ORBO™ 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50 W,W,F separators

Catalog No. 111011109
Change view
Click to view available options
Length (Metric):
75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111011109 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111011109 Supplier MilliporeSigma™ Supelco™ Supplier No. 20229

Used for gas and vapor sampling

ORBO™ 52 Small Activated Silica Gel (20/40) 150/75 mg solvent desorption tube is designed for gas and vapor sampling. It contains two beds of the same selective adsorbent separated by glass wool or foam. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Specifications

Material W,W,F Separators
Quantity 50/Pk.
Length (Metric) 75 mm
Diameter (Metric) 6 mm

ORBO is a trademark of Sigma-Aldrich Co. LLC