Learn More
Research Products Corporation Orbi-Shaker CO2 Remote Control Shaker, 12 x 13 Inch Non-Slip Platform
Numéro de catalogue. 501976821
Encompass
Orbi-Shaker CO2 performs shaking, mixing and aeration operations in any laboratory environment including CO2 incubators. Withstands temperatures from 0C to 60C and RH up to 100%. Unique remote control module allows digital setting and adjusting of shaker speed and cycle time without opening incubator door. Remote module connects to shaker with a five foot long thin ribbon cable that fits easily through an access port. On incubator models without a port, the cable slips under door gasket without interrupting chamber seal.
Supplied complete with shaker unit, non-slip aluminum platform, remote control module and ribbon cable.
Optional magnetic platforms are available separately for use with flasks and tubes.
Numéro de catalogue. 501976821 Fournisseur Research Products Corporation Code fournisseur 260120
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Orbi-Shaker CO2 performs shaking, mixing and aeration operations in any laboratory environment including CO2 incubators. Withstands temperatures from 0C to 60C and RH up to 100%. Unique remote control module allows digital setting and adjusting of shaker speed and cycle time without opening incubator door. Remote module connects to shaker with a five foot long thin ribbon cable that fits easily through an access port. On incubator models without a port, the cable slips under door gasket without interrupting chamber seal.
Supplied complete with shaker unit, non-slip aluminum platform, remote control module and ribbon cable.
Optional magnetic platforms are available separately for use with flasks and tubes.