Learn More
Electron Microscopy Sciences OPTO-WIPES SIZE 6X6IN
Numéro de catalogue. 50949333
Encompass
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily. Features super-soft fibers to prevent the scratching of precision surfaces. Strong fiber matrix maintains integrity when wet resulting in durability. Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol. Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.
Numéro de catalogue. 50949333 Fournisseur Electron Microscopy Sciences Code fournisseur 7175502
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments. This special material offers softness, is lint-free, and traps particles and residue quite easily. Features super-soft fibers to prevent the scratching of precision surfaces. Strong fiber matrix maintains integrity when wet resulting in durability. Highly absorbent by retaining and withstanding solvents such as MEK, methylene chloride, acetone, and alcohol. Particle trapping matrix lifts particles and residue away from the surface. Washable and reusable.